Assay Details
| Gene Symbol: 4930412F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015119.1 | - | 1 - 2 | 75 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
coatomer protein complex, subunit gamma 2 |
Copg2 |
AW227625, gamma-2-COP |
-105.0 | Anti-sense | Chr. 6 - 30809559 - 30896794 on NCBI Build GRCm38 |
| Downstream | Kruppel-like factor 14 | Klf14 |
5330411L03Rik, AL022736, BTEB5 |
47.8kb | Anti-sense | Chr. 6 - 30956021 - 30958990 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930412F09Rik
Genomic Map