Assay Details
| Gene Symbol: 4930414N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015140.1 | - | 1 - 2 | 365 | 96 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY714688.1 | - | 365 | Chr. - - [-] on Build | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31641 |
Gm31641 | -33.7kb | Sense | Chr. 16 - 95184537 - 95263455 on NCBI Build GRCm38 | |
| Downstream | avian erythroblastosis virus E-26 (v-ets) oncogene related | Erg | D030036I24Rik | 57.5kb | Sense | Chr. 16 - 95359169 - 95586593 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930414N08Rik
Genomic Map