Assay Details
| Gene Symbol: 4930423K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015190.1 | - | 1041 | 101 | ||
| AK086661.1 | - | 1333 | 101 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SCO cytochrome oxidase deficient homolog 2 (yeast) | Sco2 | -914.0 | Anti-sense | Chr. 15 - 89371637 - 89373818 on NCBI Build GRCm38 | |
| Downstream | outer dense fiber of sperm tails 3B | Odf3b | 2010001J22Rik | 2.0kb | Anti-sense | Chr. 15 - 89377447 - 89379681 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930423K06Rik
Genomic Map