Assay Details
| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015211.1 | - | 288 | 94 | ||
| AK019575.1 | - | 398 | 94 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | STE20-related kinase adaptor beta | Stradb |
AA792893, Als2cr2, B830008M19, D1Ucla2, ILPIP, ILPIPA, PRO1038, Papk, Syradb |
-2.0kb | Anti-sense | Chr. 1 - 58973527 - 58995126 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Als2cr11
Genomic Map