Assay Details
| Gene Symbol: 4930429C20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015232.1 | - | 1 - 2 | 247 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4251 | Gm4251 | -150.0kb | Sense | Chr. 14 - 71251273 - 71271059 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930434J06 gene | 4930434J06Rik | 435.1kb | Sense | Chr. 14 - 71856709 - 71920438 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930429C20Rik
Genomic Map