Assay Details
| Gene Symbol: 4930429E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI847387.1 | - | 95 | |||
| AK015234.1 | - | 546 | 95 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein S6 kinase polypeptide 1 | Rps6ka1 |
Mapkapk-1a, Rsk, Rsk-1, Rsk1, S6K-alpha-1, p90-Rsk1, p90Rsk1, p90S6K, p90rsk |
-23.0kb | Anti-sense | Chr. 4 - 133847290 - 133887797 on NCBI Build GRCm38 |
| Downstream | high mobility group nucleosomal binding domain 2 | Hmgn2 |
HMG-17, Hmg17 |
53.3kb | Anti-sense | Chr. 4 - 133964739 - 133967991 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930429E23Rik
Genomic Map