Assay Details
| Gene Symbol: 4930429N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015244.1 | - | 1 | 1647 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | growth associated protein 43 | Gap43 |
B-50, Basp2, GAP-43 |
-152.6kb | Sense | Chr. 16 - 42248552 - 42340651 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4932412D23 gene | 4932412D23Rik |
ENSMUSG00000075070, Gm10809 |
230.6kb | Sense | Chr. 16 - 42725702 - 42875762 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930429N05Rik
Genomic Map