Assay Details
| Gene Symbol: 4930431H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015268.1 | - | 1 | 351 | 104 | |
| Gene Symbol: Pde7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122759.2 | NP_001116231.1 | 13 | 6062 | 104 | |
| NM_008802.3 | NP_032828.2 | 13 | 5792 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial fission regulator 1 | Mtfr1 |
1300002C08Rik, 1700080D04Rik, 2810026O10Rik, 4930579E05Rik, Chppr, Fam54a2 |
-2.3kb | Anti-sense | Chr. 3 - 19187326 - 19220817 on NCBI Build GRCm38 |
| Downstream |
DnaJ (Hsp40) homolog, subfamily C, member 5 beta |
Dnajc5b | 1700008A05Rik | 284.9kb | Anti-sense | Chr. 3 - 19508591 - 19610862 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930431H11Rik
Genomic Map