Assay Details
| Gene Symbol: Dpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U48240.1 | - | 1 | 149 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein phosphatase 1, regulatory (inhibitor) subunit 14A |
Ppp1r14a |
1110001M11Rik, Cpi17 |
-14.9kb | Sense | Chr. 7 - 29289320 - 29293390 on NCBI Build GRCm38 |
| Downstream | signal-induced proliferation-associated 1 like 3 | Sipa1l3 | 2610511M17Rik | 11.9kb | Anti-sense | Chr. 7 - 29320372 - 29518943 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the U48240.1 transcripts.
This assay is in the 5'UTR region of the U48240.1 transcripts.
Set Membership: |
|

Dpf1
Genomic Map