Assay Details
| Gene Symbol: D11Bhm181e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | X81716.1 | - | 1 | 51 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 97 | Tmem97 |
1810014L12Rik, AI115531, AL022956, D11Bhm182e |
-10.7kb | Sense | Chr. 11 - 78541817 - 78550735 on NCBI Build GRCm38 |
| Downstream | nemo like kinase | Nlk | AI194375 | 5.6kb | Sense | Chr. 11 - 78567168 - 78697595 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D11Bhm181e
Genomic Map