Assay Details
| Gene Symbol: Gdap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | Y17854.1 | - | 1 | 148 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dishevelled associated activator of morphogenesis 1 | Daam1 | Sense | Chr. 12 - 71831051 - 71992376 on NCBI Build GRCm38 | ||
| Downstream | G protein-coupled receptor 135 | Gpr135 | PAFR | 220.0kb | Anti-sense | Chr. 12 - 72069618 - 72070991 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gdap5
Genomic Map