Assay Details
| Gene Symbol: Gdap7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | Y17856.1 | - | 1 | 350 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930467D21 gene | 4930467D21Rik |
4930448L09Rik, BB013444 |
-365.9kb | Anti-sense | Chr. 5 - 97392417 - 97588126 on NCBI Build GRCm38 |
| Downstream | PR domain containing 8 | Prdm8 |
4930565F05Rik, PFM5 |
212.7kb | Anti-sense | Chr. 5 - 98167198 - 98187454 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gdap7
Genomic Map