Assay Details
| Gene Symbol: Mbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF072244.1 | - | 1 | 65 | 73 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DT922271.1 | - | 227 | Chr. - - [-] on Build | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
polymerase (DNA directed), iota |
Poli | Rad30b | -63.9kb | Anti-sense | Chr. 18 - 70508680 - 70530334 on NCBI Build GRCm38 |
| Downstream | deleted in colorectal carcinoma | Dcc |
C030036D22Rik, Igdcc1 |
659.2kb | Anti-sense | Chr. 18 - 71253613 - 72351231 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mbd2
Genomic Map