Assay Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF498299.1 | - | 3 - 4 | 256 | 68 | |
| AF498300.1 | - | 3 - 4 | 256 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 370 | Mir370 |
Mirn370, mir-370, mmu-mir-370 |
-8.7kb | Sense | Chr. 12 - 109618258 - 109618336 on NCBI Build GRCm38 |
| Downstream | snoRNA AF357425 | AF357425 | MBII-48 | 1.8kb | Sense | Chr. 12 - 109636012 - 109636080 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Rian
Genomic Map