Assay Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028261.1 | 6 - 7 | 651 | 92 | ||
| GenBank mRNA | AF498301.1 | - | 3 - 4 | 248 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 370 | Mir370 |
Mirn370, mir-370, mmu-mir-370 |
-8.7kb | Sense | Chr. 12 - 109618258 - 109618336 on NCBI Build GRCm38 |
| Downstream | snoRNA AF357426 | AF357426 | MBII-49 | 2.5kb | Sense | Chr. 12 - 109642838 - 109642904 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Rian
Genomic Map