Assay Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028261.1 | 9 - 10 | 858 | 112 | ||
| GenBank mRNA | AF498301.1 | - | 6 - 7 | 455 | 112 | |
| AF498302.1 | - | 6 - 7 | 504 | 112 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | snoRNA AF357426 | AF357426 | MBII-49 | -510.0 | Sense | Chr. 12 - 109642838 - 109642904 on NCBI Build GRCm38 |
| Downstream | snoRNA DQ267100 | DQ267100 | 4.3kb | Sense | Chr. 12 - 109649617 - 109649687 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Rian
Genomic Map