Assay Details
| Gene Symbol: Kcnq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ002199.1 | - | 1 | 58 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
transient receptor potential cation channel, subfamily M, member 5 |
Trpm5 |
9430099A16Rik, LTrpC-5, Ltrpc5, Mtr1 |
-29.9kb | Anti-sense | Chr. 7 - 143071529 - 143094642 on NCBI Build GRCm38 |
| Downstream | KCNQ1 overlapping transcript 1 | Kcnq1ot1 |
Kvlqt1-as, Lit1, Tssc8 |
88.4kb | Anti-sense | Chr. 7 - 143213111 - 143296547 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kcnq1
Genomic Map