Assay Details
| Gene Symbol: 0610042B23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002901.1 | - | 1 | 57 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | translocase of outer mitochondrial membrane 20 homolog (yeast) | Tomm20 |
1810060K07Rik, BB284719, Gm19268, MAS20, MOM19, TOM20, mKIAA0016 |
-166.0 | Anti-sense | Chr. 8 - 126930664 - 126945921 on NCBI Build GRCm38 |
| Downstream | RNA binding motif protein 34 | Rbm34 |
4930547K05Rik, 6330444B11, D8Ertd233e |
695.0 | Anti-sense | Chr. 8 - 126947172 - 126971126 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

0610042B23Rik
Genomic Map