Assay Details
| Gene Symbol: Dhrs7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK003267.1 | - | 2 - 3 | 214 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 12302 | Gm12302 | OTTMUSG00000005931 | -8.7kb | Sense | Chr. 11 - 67774406 - 67789998 on NCBI Build GRCm38 |
| Downstream | ubiquitin specific peptidase 43 | Usp43 | 53.7kb | Anti-sense | Chr. 11 - 67854523 - 67922153 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Dhrs7c
Genomic Map