Assay Details
| Gene Symbol: Fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133765.4 | NP_598526.3 | 9 | 4078 | 62 | |
| GenBank mRNA | AI838208.1 | - | 1 | 62 | ||
| AK003388.1 | - | 1 | 1246 | 62 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700030M09 gene | 1700030M09Rik | -1.8kb | Anti-sense | Chr. 8 - 121544388 - 121547652 on NCBI Build GRCm38 | |
| Downstream | microtubule-associated protein 1 light chain 3 beta | Map1lc3b |
1010001C15Rik, Atg8, LC3b, Map1lc3 |
39.5kb | Anti-sense | Chr. 8 - 121590468 - 121598047 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fbxo31
Genomic Map