Assay Details
| Gene Symbol: 1110035D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI465507.1 | - | 1 | 77 | ||
| AK004106.1 | - | 1 | 150 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1191 | Mir1191 |
Mirn1191, mmu-mir-1191, mmu-mir-1191a |
-99.0 | Sense | Chr. 7 - 27205536 - 27205655 on NCBI Build GRCm38 |
| Downstream | inositol 1,4,5-trisphosphate 3-kinase C | Itpkc | 9130023N17Rik | 1.0kb | Anti-sense | Chr. 7 - 27207170 - 27228597 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1110035D15Rik
Genomic Map