Assay Details
Gene Symbol: Fendrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004221.1 | - | 2 - 3 | 503 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 26878 |
Gm26878 | -177.6kb | Anti-sense | Chr. 8 - 120879399 - 120881217 on NCBI Build GRCm38 | |
Downstream | forkhead box F1 | Foxf1 |
AI450827, FREAC1, Foxf1a, Freac-1, HFH-8, Hfh8 |
24.6kb | Anti-sense | Chr. 8 - 121084386 - 121088154 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
