Assay Details
| Gene Symbol: 1110060G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004326.1 | - | 1 | 248 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
membrane associated guanylate kinase, WW and PDZ domain containing 2 |
Magi2 |
AIP-1, Acvri1, Acvrinp1, Acvrip1, Magi-2, S-SCAM, mKIAA0705 |
-23.6kb | Sense | Chr. 5 - 19223283 - 20704792 on NCBI Build GRCm38 |
| Downstream | putative homeodomain transcription factor 2 | Phtf2 |
1110054G21Rik, 9530062N20Rik, AI447096, C77923 |
29.8kb | Anti-sense | Chr. 5 - 20758664 - 20882124 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1110060G06Rik
Genomic Map