Assay Details
| Gene Symbol: 1110064A23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004365.1 | - | 1 | 350 | 82 | |
| BC040515.1 | - | 1 | 1413 | 82 | ||
| BF180451.1 | - | 1 | 443 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | laminin gamma 3 | Lamc3 |
AI562206, AW240805 |
-2.0kb | Sense | Chr. 2 - 31887281 - 31946535 on NCBI Build GRCm38 |
| Downstream | allograft inflammatory factor 1-like | Aif1l |
2810003C17Rik, AI043124, C87647, Iba2 |
1.2kb | Sense | Chr. 2 - 31950303 - 31973442 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1110064A23Rik
Genomic Map