Assay Details
| Gene Symbol: 1110065H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004374.1 | - | 1 | 752 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
basic helix-loop-helix domain containing, class B9 |
Bhlhb9 | 2700087I09Rik | -91.6kb | Sense | Chr. X - 135841639 - 135891081 on NCBI Build GRCm38 |
| Downstream | adipocyte-related X-chromosome expressed sequence 2 | Arxes2 |
2900062L11Rik, Spc22/23, Spcs3 |
10.2kb | Sense | Chr. X - 135993820 - 135995359 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1110065H08Rik
Genomic Map