Assay Details
| Gene Symbol: 1500002J14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005119.1 | - | 1 | 650 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 8109 | Mir8109 |
Gm27495, mmu-mir-8109 |
-53.0 | Sense | Chr. 8 - 85700905 - 85701021 on NCBI Build GRCm38 |
| Downstream | integrin alpha FG-GAP repeat containing 1 | Itfg1 |
2310047C21Rik, AI314457, Cda08, D8Wsu49e |
15.7kb | Anti-sense | Chr. 8 - 85717557 - 85840949 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1500002J14Rik
Genomic Map