Assay Details
| Gene Symbol: 1500005C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005156.1 | - | 1 | 2247 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurogenin 2 | Neurog2 |
Atoh4, Math4A, bHLHa8, ngn-2, ngn2 |
-79.0kb | Anti-sense | Chr. 3 - 127633145 - 127635631 on NCBI Build GRCm38 |
| Downstream | TRAF-interacting protein with forkhead-associated domain | Tifa | T2bp | 71.8kb | Anti-sense | Chr. 3 - 127788857 - 127798394 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1500005C15Rik
Genomic Map