Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500015156.mm.1 |
TC0500001650.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015156.mm.1 |
NR_045821.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015156.mm.1 |
NR_045821.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015159.mm.1 |
TC0500001650.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015159.mm.1 |
NR_045821.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015159.mm.1 |
NR_045821.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015145.mm.1 |
TC0500001650.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015145.mm.1 |
NR_045821.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015145.mm.1 |
NR_045821.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015158.mm.1 |
TC0500001650.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015158.mm.1 |
NR_045821.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015158.mm.1 |
NR_045821.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015157.mm.1 |
TC0500001650.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015157.mm.1 |
NR_045821.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015157.mm.1 |
NR_045821.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015152.mm.1 |
TC0500001650.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500015152.mm.1 |
NR_045821.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500015152.mm.1 |
NR_045821.1 |
Refseq |
Murine Genome U74v2 Set |
133149_i_at |
- |
- |
Murine Genome U74v2 Set |
167882_PM_at |
- |
- |
Murine Genome U74v2 Set |
133150_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
133149_PM_i_at |
- |
- |
Murine Genome U74v2 Set |
133150_f_at |
- |
- |
Murine Genome U74v2 Set |
167882_at |
- |
- |