Assay Details
| Gene Symbol: 9130025I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018650.1 | - | 1 | 655 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger and WD repeat domain 2 | Rfwd2 |
AI316802, C80879, Cop1 |
Sense | Chr. 1 - 159232174 - 159347638 on NCBI Build GRCm38 | |
| Downstream |
POU domain, class 5, transcription factor 1 pseudogene |
Gm21053 |
Oct4, Pou5f1 |
4.0kb | Sense | Chr. 1 - 159293805 - 159294189 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9130025I19Rik
Genomic Map