Assay Details
Gene Symbol: 9130401L11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK018654.1 | - | 1 | 1951 | 65 | |
AK036628.1 | - | 1 | 1790 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | forkhead box P1 | Foxp1 |
3110052D19Rik, 4932443N09Rik, AI461938, AW494214 |
Sense | Chr. 6 - 98925342 - 99523024 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 19665 |
Gm19665 | 125.9kb | Sense | Chr. 6 - 99097669 - 99108316 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
