Assay Details
| Gene Symbol: 9130403I23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018659.1 | - | 1 | 1354 | 69 | |
| AK034835.1 | - | 1 | 1381 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosomal protein S28 | Mrps28 |
1500012D08Rik, AW061224 |
-1.8kb | Anti-sense | Chr. 3 - 8802146 - 8923857 on NCBI Build GRCm38 |
| Downstream | tumor protein D52 | Tpd52 |
AI043038, mD52 |
1.5kb | Anti-sense | Chr. 3 - 8928626 - 9004769 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9130403I23Rik
Genomic Map