Assay Details
| Gene Symbol: 1700052M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018857.1 | - | 1 | 150 | 64 | |
| AV046246.3 | - | 1 | 166 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 1564 | Gm1564 | Gm663 | Anti-sense | Chr. 11 - 102664966 - 102682240 on NCBI Build GRCm38 | |
| Downstream | coiled-coil domain containing 43 | Ccdc43 |
2610001E01Rik, D11Ertd707e |
19.4kb | Sense | Chr. 11 - 102684684 - 102697943 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700052M18Rik
Genomic Map