Assay Details
| Gene Symbol: 1700089I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018915.1 | - | 1 | 251 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | charged multivesicular body protein 4B | Chmp4b |
2010012F05Rik, C76846, Snf7-2 |
-87.5kb | Sense | Chr. 2 - 154657026 - 154694783 on NCBI Build GRCm38 |
| Downstream | hnRNP-associated with lethal yellow | Raly |
AI663842, Merc |
8.4kb | Sense | Chr. 2 - 154791097 - 154871010 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700089I07Rik
Genomic Map