Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1456546_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1436419_PM_a_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1455692_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1436419_PM_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1436419_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456546_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1455692_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456546_at |
- |
- |
Mouse Genome 430 2.0 Array |
1455692_PM_x_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
67522_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
67522_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1700016639.mm.1 |
NR_045287.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016639.mm.1 |
NR_045287.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016639.mm.1 |
TC1700001806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016637.mm.1 |
NR_045287.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016637.mm.1 |
NR_045287.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016637.mm.1 |
TC1700001806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016636.mm.1 |
NR_045287.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016636.mm.1 |
NR_045287.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016636.mm.1 |
TC1700001806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016638.mm.1 |
NR_045287.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016638.mm.1 |
NR_045287.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016638.mm.1 |
TC1700001806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016640.mm.1 |
NR_045287.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016640.mm.1 |
NR_045287.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016640.mm.1 |
TC1700001806.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1456546_at |
- |
- |
Mouse Expression Set 430 |
1436419_PM_a_at |
- |
- |
Mouse Expression Set 430 |
1436419_a_at |
- |
- |
Mouse Expression Set 430 |
1455692_x_at |
- |
- |
Mouse Expression Set 430 |
1455692_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1456546_PM_at |
- |
- |
Murine Genome U74v2 Set |
161004_at |
- |
- |
Murine Genome U74v2 Set |
161004_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1456546_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1436419_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1455692_PM_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1436419_PM_a_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1456546_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1455692_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1436419_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1436419_PM_a_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1456546_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1455692_PM_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1456546_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1455692_x_at |
- |
- |