Assay Details
| Gene Symbol: 1700040B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018986.1 | - | 1 | 51 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dysbindin (dystrobrevin binding protein 1) domain containing 1 | Dbndd1 |
2810427I04Rik, D8Ertd590e |
-308.0 | Sense | Chr. 8 - 123504718 - 123518041 on NCBI Build GRCm38 |
| Downstream | growth arrest specific 8 | Gas8 | Gas11 | 46.0 | Anti-sense | Chr. 8 - 123518835 - 123536657 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700040B14Rik
Genomic Map