Assay Details
| Gene Symbol: 1700129O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018999.1 | - | 1 | 446 | 77 | |
| BM942913.1 | - | 1 | 321 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
FYVE, RhoGEF and PH domain containing 3 |
Fgd3 |
5830461L01Rik, ZFYVE5 |
-20.4kb | Sense | Chr. 13 - 49263110 - 49320464 on NCBI Build GRCm38 |
| Downstream | bicaudal D homolog 2 (Drosophila) | Bicd2 | 173.0 | Anti-sense | Chr. 13 - 49341549 - 49387026 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700129O19Rik
Genomic Map