Assay Details
| Gene Symbol: 1810056I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019035.1 | - | 1 | 154 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tryptophanyl-tRNA synthetase | Wars |
TrpRS, WRS |
Sense | Chr. 12 - 108860030 - 108894174 on NCBI Build GRCm38 | |
| Downstream | WD repeat domain 25 | Wdr25 | B930090D16Rik | 719.0 | Anti-sense | Chr. 12 - 108894228 - 109028452 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1810056I18Rik
Genomic Map