Assay Details
| Gene Symbol: Ccdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019067.1 | - | 61 | 125 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
eukaryotic translation initiation factor 3, subunit M |
Eif3m |
Ga17, Pcid1, Tango7 |
-94.0 | Anti-sense | Chr. 2 - 104999656 - 105017027 on NCBI Build GRCm38 |
| Downstream |
Wilms tumor 1 homolog, opposite strand |
Wt1os |
6030498F17, AI314831, Wt1-as |
58.5kb | Anti-sense | Chr. 2 - 105076538 - 105126510 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc73
Genomic Map