Assay Details
| Gene Symbol: 2610007O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011339.1 | - | 1 | 352 | 72 | |
| AK019189.1 | - | 1 | 352 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | maestro heat-like repeat family member 8 | Mroh8 | 4922505G16Rik | -45.0kb | Anti-sense | Chr. 2 - 157208548 - 157279549 on NCBI Build GRCm38 |
| Downstream | growth hormone releasing hormone | Ghrh | Ghrf | 4.3kb | Anti-sense | Chr. 2 - 157329496 - 157346736 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2610007O09Rik
Genomic Map