Assay Details
| Gene Symbol: Maf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019255.1 | - | 50 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream | dynein light chain roadblock-type 2 | Dynlrb2 |
1700009A04Rik, DNLC2B, Dncl2b |
797.2kb | Anti-sense | Chr. 8 - 116505015 - 116515915 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Maf
Genomic Map