Assay Details
| Gene Symbol: Hnrnpab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019387.1 | - | 252 | 91 | ||
| AK047205.1 | - | 2067 | 91 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
collagen, type XXIII, alpha 1 |
Col23a1 |
2810458L13Rik, AI429655 |
-13.2kb | Sense | Chr. 11 - 51289678 - 51583925 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpab
Genomic Map