Assay Details
| Gene Symbol: 3010031K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019402.1 | - | 1 | 656 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DNA segment, Chr 8, ERATO Doi 82, expressed |
D8Ertd82e |
9830148H23, NACK, Sgk223, mFLJ00269 |
Sense | Chr. 8 - 36094625 - 36147787 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 38414 |
Gm38414 | A230084N10 | 59.0kb | Sense | Chr. 8 - 36156010 - 36161582 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

3010031K01Rik
Genomic Map