Assay Details
| Gene Symbol: 3110033O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019423.1 | - | 1 | 51 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lin-7 homolog A (C. elegans) | Lin7a |
AI848705, LIN-7A, MALS-1, TIP-33, Veli, Veli1 |
Sense | Chr. 10 - 107271798 - 107425146 on NCBI Build GRCm38 | |
| Downstream | myogenic factor 5 | Myf5 |
B130010J22Rik, Myf-5, bHLHc2 |
147.0kb | Anti-sense | Chr. 10 - 107482908 - 107486927 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

3110033O22Rik
Genomic Map