Assay Details
| Gene Symbol: 4930420G21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019579.1 | - | 1 | 249 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 33255 |
Gm33255 | -44.6kb | Anti-sense | Chr. 16 - 88352647 - 88381817 on NCBI Build GRCm38 | |
| Downstream | claudin 17 | Cldn17 | 78.4kb | Anti-sense | Chr. 16 - 88505807 - 88506978 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930420G21Rik
Genomic Map