Assay Details
Gene Symbol: 4930428D20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019586.1 | - | 1 - 2 | 73 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cell adhesion molecule 2 | Cadm2 |
2900078E11Rik, 9330131D06, A830029E02Rik, Igdf4d, Igsf4d, NECL3, SynCAM 2, SynCAM2 |
-2.2mb | Sense | Chr. 16 - 66655421 - 67621200 on NCBI Build GRCm38 |
Downstream | spermatogenesis associated glutamate (E)-rich protein 2 | Speer2 | SPEER-2 | 56.5kb | Sense | Chr. 16 - 69856874 - 69863794 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
