Assay Details
| Gene Symbol: 4930486F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038024.1 | 2 - 3 | 161 | 98 | ||
| GenBank mRNA | AK019657.1 | - | 2 - 3 | 157 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synapsin III | Syn3 | Anti-sense | Chr. 10 - 86048746 - 86499019 on NCBI Build GRCm38 | ||
| Downstream | tissue inhibitor of metalloproteinase 3 | Timp3 | Timp-3 | 186.8kb | Sense | Chr. 10 - 86300412 - 86349505 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930486F22Rik
Genomic Map