Assay Details
| Gene Symbol: 4930527L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019705.1 | - | 1 | 146 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 26B | Kif26b |
4832420M10, BC056349, D230039L06Rik |
Anti-sense | Chr. 1 - 178525741 - 178939200 on NCBI Build GRCm38 | |
| Downstream | SET and MYND domain containing 3 | Smyd3 |
2410008A19Rik, Zmynd1 |
363.1kb | Sense | Chr. 1 - 178951960 - 179518052 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930527L19Rik
Genomic Map