Assay Details
| Gene Symbol: Tac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019709.1 | - | 137 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 19272 |
Gm19272 | -182.9kb | Sense | Chr. 6 - 7301886 - 7379505 on NCBI Build GRCm38 | |
| Downstream | asparagine synthetase | Asns | 110.9kb | Anti-sense | Chr. 6 - 7675165 - 7693182 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Tac1
Genomic Map