Assay Details
| Gene Symbol: 4930539C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019724.1 | - | 1 | 343 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BMP-binding endothelial regulator | Bmper |
3110056H04Rik, CV-2, Crim3, Cv2 |
-264.7kb | Sense | Chr. 9 - 23223076 - 23485215 on NCBI Build GRCm38 |
| Downstream | neuropeptide S receptor 1 | Npsr1 |
9330128H10Rik, GPRA, Gpr154, MVTR, PGR14, VRR1 |
347.6kb | Sense | Chr. 9 - 24097950 - 24317590 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930539C01Rik
Genomic Map