Assay Details
| Gene Symbol: 4930589O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019834.1 | - | 643 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UDP-glucose dehydrogenase | Ugdh | Udpgdh | -57.2kb | Anti-sense | Chr. 5 - 65413207 - 65435842 on NCBI Build GRCm38 |
| Downstream | ubiquitin-conjugating enzyme E2K | Ube2k |
AW492011, D5Ertd601e, E2-25k, Hip2, Hypg, Lig |
43.3kb | Sense | Chr. 5 - 65537238 - 65598989 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930589O11Rik
Genomic Map